Gene Transcripts

Gene Symbol
Nlrp5
Entrez Gene ID
23968
Gene Name
NLR family, pyrin domain containing 5
Gene Aliases
Mater, Nalp5, Op1, PAN11
Chromosome Location
Chr. 7: 23385897 - 23441921 on Build GRCm38
UniGene ID
Mm.333653
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF074018.1 - 2 112
AK087774.1 - 3 180
AY196361.1 - 2 112
AY196362.1 - 2 112
AY329484.1 - 2 112
AY329485.1 - 2 112
AY329486.1 - 2 112
AY329487.1 - 2 112
AY329488.1 - 2 112
AY329489.1 - 2 112
AY329490.1 - 2 112
AY329491.1 - 2 112
BC053384.1 - 3 179 30049556

View Full Product Details