Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290707.1 | NP_001277636.1 | 23 | 3184 | |
NM_011866.2 | NP_035996.2 | 22 | 3138 | ||
GenBank mRNA | AF110507.1 | - | 22 | 3009 | |
AK039249.1 | - | 23 | 3184 | ||
AK147690.1 | - | 22 | 3056 | ||
AK162221.1 | - | 22 | 2793 | ||
AK162804.1 | - | 20 | 2679 | ||
AK166310.1 | - | 14 | 2150 | ||
AY360383.1 | - | 22 | 3138 | ||
BC113201.1 | - | 22 | 3004 | 40048238 |