Gene Transcripts

Gene Symbol
Eif2ak4
Entrez Gene ID
27103
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 4
Gene Aliases
2610011M03, GCN2, MGCN2
Chromosome Location
Chr. 2: 118388578 - 118475234 on Build GRCm38
UniGene ID
Mm.217616
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177806.1 NP_001171277.1 3, 4 309
NM_013719.3 NP_038747.2 4, 5 613
GenBank mRNA AF193343.1 - 4, 5 605
AF193344.1 - 4, 5 697
AJ243533.1 - 4, 5 613
AK013758.1 - 4, 5 569
BC023958.1 - 4, 5 636
BC072637.1 - 3, 4 309 30354433

View Full Product Details