Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013745.5 | NP_038773.1 | 8 | 1072 | |
GenBank mRNA | AF159549.1 | - | 8 | 1030 | |
AK032324.1 | - | 8 | 1073 | ||
AK156205.1 | - | 8 | 1059 | ||
BC020364.1 | - | 8 | 1047 | ||
BC056192.1 | - | 8 | 1058 | 30024494 |