Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199004.1 | NP_001185933.1 | 2 | 464 | |
NM_013747.4 | NP_038775.1 | 2 | 306 | ||
GenBank mRNA | AB016784.1 | - | 2 | 465 | |
AF026274.1 | - | 2 | 296 | ||
AK007592.1 | - | 2 | 311 | ||
AK138455.1 | - | 2 | 465 | ||
AK152533.1 | - | 2 | 314 | ||
AK153010.1 | - | 2 | 314 | ||
AK168765.1 | - | 2 | 310 | ||
BC016883.1 | - | 2 | 1034 | 4235952 | |
BC086782.1 | - | 2 | 306 | 6813878 |