Gene Transcripts

Gene Symbol
ADRM1
Entrez Gene ID
11047
Gene Name
adhesion regulating molecule 1
Gene Aliases
ARM-1, ARM1, GP110
Chromosome Location
Chr. 20: 62302056 - 62308862 on Build GRCh38
UniGene ID
Hs.90107
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007002.3 NP_008933.2 3 438
NM_175573.2 NP_783163.1 3 434
XM_005260257.1 XP_005260314.1 4 525
XM_011528503.1 XP_011526805.1 3 1273
XM_017027602.1 XP_016883091.1 3 1273
XM_017027603.1 XP_016883092.1 4 525
XM_017027604.1 XP_016883093.1 3 1269
XM_017027605.1 XP_016883094.1 3 1273
GenBank mRNA AK297567.1 - 3 332
AK307459.1 - 368
AL558522.3 - 3 370
BC017245.2 - 3 334 4890786
CR749213.1 - 1 1055
D64154.1 - 3 329

View Full Product Details