Gene Transcripts

Gene Symbol
SRSF1
Entrez Gene ID
6426
Gene Name
serine and arginine rich splicing factor 1
Gene Aliases
ASF, SF2, SF2p33, SFRS1, SRp30a
Chromosome Location
Chr. 17: 58000919 - 58007346 on Build GRCh38
UniGene ID
Hs.68714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001078166.1 NP_001071634.1 3 1972
NM_006924.4 NP_008855.1 4 1772
GenBank mRNA AB062124.1 - 4 1653
AB209558.1 - 3 1857
AK298527.1 - 3 1354
BC010264.1 - 4 1598 2900101

View Full Product Details