Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053403.2 | NP_445855.1 | 8, 9 | 1377 | |
XM_006247432.2 | XP_006247494.1 | 8, 9 | 3346 | ||
XM_006247433.2 | XP_006247495.1 | 8, 9 | 1229 | ||
XM_006247434.2 | XP_006247496.1 | 8, 9 | 1629 | ||
XM_008768125.1 | XP_008766347.1 | 8, 9 | 1163 | ||
GenBank mRNA | AF190121.1 | - | 8, 9 | 978 | |
BC081757.1 | - | 8, 9 | 1377 | 7109771 |