Gene Transcripts

Gene Symbol
Ctsd
Entrez Gene ID
171293
Gene Name
cathepsin D
Gene Aliases
Chromosome Location
Chr. 1: 215541570 - 215553446 on Build Rnor_6.0
UniGene ID
Rn.11085
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134334.2 NP_599161.2 2 254
GenBank mRNA BC062032.1 - 2 254
X54467.1 - 2 199

View Full Product Details