Gene Transcripts

Gene Symbol
Kcnip4
Entrez Gene ID
259243
Gene Name
Kv channel interacting protein 4
Gene Aliases
Kchip4
Chromosome Location
Chr. 14: 65549366 - 66747926 on Build Rnor_6.0
UniGene ID
Rn.205864
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181365.2 NP_852030.1 1160
GenBank mRNA AF345444.1 - 1160
AF345445.1 - 7 730

View Full Product Details