Gene Transcripts

Gene Symbol
Chn1
Entrez Gene ID
84030
Gene Name
chimerin 1
Gene Aliases
Chromosome Location
Chr. 3: 60512360 - 60668281 on Build Rnor_6.0
UniGene ID
Rn.11166
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032083.1 NP_114472.1 1 524
GenBank mRNA BC094519.1 - 1 573
X67250.1 - 1 524

View Full Product Details