Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193640.1 | NP_001180569.1 | 2 | 315 | |
NM_001257965.1 | NP_001244894.1 | 4 | 238 | ||
NM_001257966.1 | NP_001244895.1 | 2 | 315 | ||
NM_201253.2 | NP_957705.1 | 2 | 315 | ||
NR_047563.1 | 2 | 315 | |||
NR_047564.1 | 2 | 315 | |||
XM_011509365.2 | XP_011507667.1 | 2 | 240 | ||
XM_011509367.1 | XP_011507669.1 | 2 | 240 | ||
XM_017000851.1 | XP_016856340.1 | 2 | 147 | ||
XM_017000852.1 | XP_016856341.1 | 2 | 240 | ||
GenBank mRNA | AB208923.1 | - | 2 | 243 | |
AF154671.1 | - | 2 | 241 | ||
AJ748821.1 | - | 2 | 106 | ||
AK289733.1 | - | 2 | 260 | ||
AK299368.1 | - | 2 | 279 | ||
AK302826.1 | - | 4 | 238 | ||
AK307844.1 | - | 2 | 258 | ||
AY043324.1 | - | 2 | 241 | ||
AY043325.1 | - | 2 | 241 | ||
BC136271.1 | - | 2 | 154 | 9020258 | |
BC143726.1 | - | 2 | 154 | ||
DC370545.1 | - | 2 | 317 | ||
DC370777.1 | - | 2 | 311 |