Gene Transcripts
- Gene Symbol
- ECD
- Entrez Gene ID
- 11319
- Gene Name
- ecdysoneless cell cycle regulator
- Gene Aliases
- GCR2, HSGT1, SGT1
- Chromosome Location
- Chr. 10: 73134524 - 73168095 on Build GRCh38
- UniGene ID
- Hs.631822
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135752.1 |
NP_001129224.1
|
2, 3 |
456 |
|
NM_001135753.1 |
NP_001129225.1
|
2, 3 |
456 |
|
NM_007265.2 |
NP_009196.1
|
2, 3 |
456 |
|
GenBank mRNA |
AK022616.1 |
- |
2, 3 |
404 |
|
AK225519.1 |
- |
2, 3 |
432 |
|
AK315711.1 |
- |
2, 3 |
429 |
|
BC000721.2 |
- |
2, 3 |
420 |
3504956 |
CR457407.1 |
- |
1, 2 |
213 |
|
D88208.1 |
- |
2, 3 |
420 |
|
DA162516.1 |
- |
2, 3 |
432 |
|
View Full Product Details