Gene Transcripts

Gene Symbol
Apex1
Entrez Gene ID
79116
Gene Name
APEX nuclease (multifunctional DNA repair enzyme) 1
Gene Aliases
APE, Apex, REF-1
Chromosome Location
Chr. 15: 27849943 - 27852083 on Build Rnor_6.0
UniGene ID
Rn.5949
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024148.1 NP_077062.1 3 253
XM_006251913.1 XP_006251975.1 3 294
GenBank mRNA AF309114.1 - 2 169
AF311054.1 - 2 131
BC078816.1 - 3 261 7127205
D44495.1 - 3 253
FQ220521.1 - 3 255
L27076.1 - 3 237

View Full Product Details