Gene Transcripts

Gene Symbol
Rab26
Entrez Gene ID
171111
Gene Name
RAB26, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 10: 13894271 - 13898578 on Build Rnor_6.0
UniGene ID
Rn.10975
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133580.1 NP_598264.1 8 644
GenBank mRNA BC061984.1 - 9 952
U18771.1 - 8 644

View Full Product Details