Gene Transcripts

Gene Symbol
LRRC29
Entrez Gene ID
26231
Gene Name
leucine rich repeat containing 29
Gene Aliases
FBL9, FBXL9
Chromosome Location
Chr. 16: 67207139 - 67227062 on Build GRCh38
UniGene ID
Hs.461000
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004055.1 NP_001004055.1 4, 5 553
NM_012163.2 NP_036295.1 4, 5 718
XM_017023126.1 XP_016878615.1 4, 5 982
XM_017023127.1 XP_016878616.1 4, 5 1233
XM_017023128.1 XP_016878617.1 4, 5 1038
XM_017023129.1 XP_016878618.1 4, 5 2124
XM_017023130.1 XP_016878619.1 4, 5 706
XM_017023131.1 XP_016878620.1 4, 5 565
XM_017023132.1 XP_016878621.1 4, 5 868
XM_017023133.1 XP_016878622.1 4, 5 894
XM_017023134.1 XP_016878623.1 4, 5 673
GenBank mRNA AF176701.1 - 4, 5 563
AK096101.1 - 2, 3 1190
AK316602.1 - 4, 5 566
BC018785.1 - 4, 5 718 4875646
BI085522.1 - 3, 4 436

View Full Product Details