Gene Transcripts
- Gene Symbol
- Grasp
- Entrez Gene ID
- 192254
- Gene Name
- GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein
- Gene Aliases
- Chromosome Location
- Chr. 7: 142869785 - 142877551 on Build Rnor_6.0
- UniGene ID
- Rn.229849
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138894.1 |
NP_620249.1
|
6, 7 |
680 |
|
GenBank mRNA |
AF374272.1 |
- |
6, 7 |
680 |
|
View Full Product Details