Gene Transcripts

Gene Symbol
Ermp1
Entrez Gene ID
373544
Gene Name
endoplasmic reticulum metallopeptidase 1
Gene Aliases
Fxna
Chromosome Location
Chr. 1: 247784830 - 247821772 on Build Rnor_6.0
UniGene ID
Rn.99609
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_184050.2 NP_908939.2 1, 2 345
XM_006231241.2 XP_006231303.1 1, 2 389
GenBank mRNA AY364634.2 - 1, 2 345

View Full Product Details