Gene Transcripts

Gene Symbol
Kcnc1
Entrez Gene ID
25327
Gene Name
potassium channel, voltage gated Shaw-related subfamily C, member 1
Gene Aliases
KShIIIB, Kv3.1, Kv4, NGK2-KV4
Chromosome Location
Chr. 1: 102414625 - 102456411 on Build Rnor_6.0
UniGene ID
Rn.33095
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012856.1 NP_036988.1 2 2305
GenBank mRNA M68880.1 - 2 2305
X62840.1 - 2 1430

View Full Product Details