Gene Transcripts

Gene Symbol
Dlx5
Entrez Gene ID
25431
Gene Name
distal-less homeobox 5
Gene Aliases
RDLX
Chromosome Location
Chr. 4: 32387741 - 32392007 on Build Rnor_6.0
UniGene ID
Rn.11248
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012943.1 NP_037075.1 3 1330
GenBank mRNA D31734.1 - 3 1327
L24443.1 - 3 1330

View Full Product Details