Gene Transcripts

Gene Symbol
Hrh2
Entrez Gene ID
25461
Gene Name
histamine receptor H 2
Gene Aliases
H2R
Chromosome Location
Chr. 17: 10912993 - 10931368 on Build Rnor_6.0
UniGene ID
Rn.138113
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012965.3 NP_037097.3 1 295
XM_008771486.1 XP_008769708.1 1 1602

View Full Product Details