Gene Transcripts

Gene Symbol
Ptgds
Entrez Gene ID
25526
Gene Name
prostaglandin D2 synthase (brain)
Gene Aliases
PH2DISO
Chromosome Location
Chr. 3: 2686125 - 2689059 on Build Rnor_6.0
UniGene ID
Rn.11400
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013015.2 NP_037147.1 2 222
GenBank mRNA FQ214359.1 - 1 474
J04488.1 - 2 222

View Full Product Details