Gene Transcripts

Gene Symbol
Rab3a
Entrez Gene ID
25531
Gene Name
RAB3A, member RAS oncogene family
Gene Aliases
RAB3
Chromosome Location
Chr. 16: 20435098 - 20439206 on Build Rnor_6.0
UniGene ID
Rn.44409
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013018.2 NP_037150.2 5 659
GenBank mRNA BC087580.1 - 5 659
FQ214415.1 - 5 627
X06889.1 - 5 631

View Full Product Details