Gene Transcripts

Gene Symbol
Hspa5
Entrez Gene ID
25617
Gene Name
heat shock protein 5
Gene Aliases
BIP, GRP78
Chromosome Location
Chr. 3: 13838304 - 13842763 on Build Rnor_6.0
UniGene ID
Rn.11088
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013083.2 NP_037215.1 3 490
GenBank mRNA BC062017.1 - 3 450
CB578015.1 - 3 490
M14050.1 - 2 366

View Full Product Details