Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242854.1 | NP_001229783.1 | 2 | 367 | |
NM_012402.3 | NP_036534.1 | 2 | 327 | ||
XM_005252840.4 | XP_005252897.1 | 2 | 231 | ||
GenBank mRNA | AK026390.1 | - | 2 | 159 | |
AK300855.1 | - | 2 | 228 | ||
AK301972.1 | - | 2 | 334 | ||
AK309198.1 | - | 2 | 231 | ||
AK310833.1 | - | 2 | 186 | ||
AK311086.1 | - | 2 | 96 | ||
AK311655.1 | - | 3 | 204 | ||
BC000392.2 | - | 2 | 149 | 2821666 | |
BM928413.1 | - | 294 | |||
DB099222.1 | - | 2 | 327 | ||
U52522.1 | - | 2 | 143 |