Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031641.1 | NP_113829.1 | 5 | 579 | |
XM_006242114.2 | XP_006242176.1 | 5 | 1371 | ||
GenBank mRNA | AF176343.1 | - | 2 | 169 | |
AF188699.1 | - | 5 | 579 | ||
DQ897974.1 | - | 1 | 27 |