Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031648.2 | NP_113836.2 | 5, 6 | 289 | |
XM_006228838.2 | XP_006228900.1 | 6, 7 | 509 | ||
XM_006228839.2 | XP_006228901.1 | 5, 6 | 284 | ||
XM_008759184.1 | XP_008757406.1 | 5, 6 | 298 | ||
GenBank mRNA | CB712570.1 | - | 5, 6 | 289 | |
FQ211349.1 | - | 5, 6 | 300 | ||
FQ224131.1 | - | 5, 6 | 252 | ||
FQ224338.1 | - | 5, 6 | 335 | ||
U72246.1 | - | 4, 5 | 203 |