Gene Transcripts

Gene Symbol
KIN
Entrez Gene ID
22944
Gene Name
Kin17 DNA and RNA binding protein
Gene Aliases
BTCD, KIN17, Rts2
Chromosome Location
Chr. 10: 7750962 - 7788027 on Build GRCh38
UniGene ID
Hs.397918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012311.3 NP_036443.1 4 365
NR_045609.1 4 365
XM_006717434.3 XP_006717497.1 4 360
XM_006717435.3 XP_006717498.1 3 233
XM_011519427.1 XP_011517729.1 4 360
XM_017015946.1 XP_016871435.1 3 236
GenBank mRNA AJ005273.1 - 4 336
AK296637.1 - 3 233
AK301789.1 - 4 365
AK309059.1 - 4 301
BC017309.2 - 4 282 5089578

View Full Product Details