Gene Transcripts

Gene Symbol
Chga
Entrez Gene ID
24258
Gene Name
chromogranin A (parathyroid secretory protein 1)
Gene Aliases
Chromosome Location
Chr. 6: 126434226 - 126445569 on Build Rnor_6.0
UniGene ID
Rn.41024
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021655.2 NP_067687.2 5, 6 549
GenBank mRNA AF145445.1 - 5, 6 402
BC087703.1 - 5, 6 529 7313842
X06832.1 - 5, 6 537

View Full Product Details