Gene Transcripts
- Gene Symbol
- Kap
- Entrez Gene ID
- 24937
- Gene Name
- kidney androgen regulated protein
- Gene Aliases
- Chromosome Location
- Chr. 4: 167549922 - 167552973 on Build Rnor_6.0
- UniGene ID
- Rn.11143
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_052802.1 |
NP_434689.1
|
2, 3 |
392 |
|
GenBank mRNA |
U25808.1 |
- |
2, 3 |
392 |
|
View Full Product Details