Gene Transcripts
- Gene Symbol
- Rbm10
- Entrez Gene ID
- 64510
- Gene Name
- RNA binding motif protein 10
- Gene Aliases
- Chromosome Location
- Chr. X: 1754869 - 1786973 on Build Rnor_6.0
- UniGene ID
- Rn.8822
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152861.4 |
NP_690600.1
|
20 |
2525 |
|
GenBank mRNA |
D83948.1 |
- |
20 |
2525 |
|
View Full Product Details