Gene Transcripts

Gene Symbol
Rnase6
Entrez Gene ID
305842
Gene Name
ribonuclease, RNase A family, 6
Gene Aliases
Chromosome Location
Chr. 15: 28080886 - 28084301 on Build Rnor_6.0
UniGene ID
Rn.230067
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206815.1 NP_996538.1 1 211
XM_006251871.2 XP_006251933.1 2 280
XM_006251872.2 XP_006251934.1 2 579
XM_008770517.1 XP_008768739.1 3 424
XM_008770518.1 XP_008768740.1 3 398
XM_008770519.1 XP_008768741.1 2 298
XM_008770520.1 XP_008768742.1 2 277

Gene Symbol
LOC103694852
Entrez Gene ID
103694852
Gene Name
ribonuclease K6-like
Gene Aliases
Chromosome Location
Chr. Un|NW_007906778: 48178 - 52172 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773787.1 XP_008772009.1 2 280
XM_008773788.1 XP_008772010.1 2 579
XM_008773789.1 XP_008772011.1 3 427
XM_008773790.1 XP_008772012.1 3 401
XM_008773791.1 XP_008772013.1 2 301
XM_008773792.1 XP_008772014.1 2 277

View Full Product Details