Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138316.3 | NP_612189.2 | 5 | 1040 | |
NM_148977.2 | NP_683878.1 | 6 | 1995 | ||
NM_148978.2 | NP_683879.1 | 6 | 1217 | ||
XM_017016333.1 | XP_016871822.1 | 7 | 1963 | ||
XM_017016334.1 | XP_016871823.1 | 7 | 1925 | ||
XM_017016335.1 | XP_016871824.1 | 7 | 2753 | ||
XM_017016336.1 | XP_016871825.1 | 6 | 1142 | ||
XM_017016337.1 | XP_016871826.1 | 7 | 2659 | ||
XM_017016338.1 | XP_016871827.1 | 4 | 1359 | ||
GenBank mRNA | AF355198.1 | - | 5 | 1024 | |
AK125428.1 | - | 7 | 1574 | ||
AK127777.1 | - | 5 | 916 | ||
AY027661.1 | - | 6 | 1995 | ||
AY027662.1 | - | 6 | 1217 | ||
AY027663.1 | - | 7 | 1311 | ||
BC026296.1 | - | 3 | 350 |