Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138316.3 | NP_612189.2 | 6 | 1293 | |
NM_148977.2 | NP_683878.1 | 7 | 2248 | ||
NM_148978.2 | NP_683879.1 | 7 | 1470 | ||
XM_017016333.1 | XP_016871822.1 | 8 | 2216 | ||
XM_017016334.1 | XP_016871823.1 | 8 | 2178 | ||
XM_017016335.1 | XP_016871824.1 | 8 | 3006 | ||
XM_017016336.1 | XP_016871825.1 | 7 | 1395 | ||
XM_017016337.1 | XP_016871826.1 | 8 | 2912 | ||
XM_017016338.1 | XP_016871827.1 | 5 | 1612 | ||
GenBank mRNA | AF355198.1 | - | 6 | 1277 | |
AK125428.1 | - | 8 | 1827 | ||
AK127777.1 | - | 6 | 1169 | ||
AY027661.1 | - | 7 | 2248 | ||
AY027662.1 | - | 7 | 1470 | ||
AY027663.1 | - | 8 | 1564 | ||
BC026296.1 | - | 4 | 603 |