Gene Transcripts

Gene Symbol
CHRM1
Entrez Gene ID
1128
Gene Name
cholinergic receptor muscarinic 1
Gene Aliases
HM1, M1, M1R
Chromosome Location
Chr. 11: 62908675 - 62921861 on Build GRCh38
UniGene ID
Hs.632119
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000738.2 NP_000729.2 2 2387
XM_011544742.2 XP_011543044.1 2 2088
GenBank mRNA BC007740.2 - 2 2108
BC022984.1 - 2 2387 4931283

View Full Product Details