Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013283.4 | NP_037415.1 | 2 | 374 | |
NM_182796.2 | NP_877725.1 | 2 | 513 | ||
GenBank mRNA | AB073390.1 | - | 2 | 288 | |
AF113225.1 | - | 2 | 170 | ||
AF182814.1 | - | 2 | 215 | ||
AJ243721.1 | - | 2 | 353 | ||
AK096178.1 | - | 2 | 276 | ||
AK291919.1 | - | 2 | 324 | ||
AK294069.1 | - | 2 | 186 | ||
AK312365.1 | - | 2 | 323 | ||
AL136664.1 | - | 2 | 287 | ||
AU279489.1 | - | 2 | 323 | ||
AW966728.1 | - | 2 | 352 | ||
AY358695.1 | - | 2 | 268 | ||
BC005218.1 | - | 2 | 294 | 3931953 | |
BC066645.1 | - | 2 | 249 | 5749043 | |
BC093030.1 | - | 2 | 283 | 30527606 | |
DA146198.1 | - | 2 | 377 | ||
DQ395260.1 | - | 3 | 355 | ||
DQ413183.1 | - | 2 | 352 |