Gene Transcripts

Gene Symbol
TONSL
Entrez Gene ID
4796
Gene Name
tonsoku-like, DNA repair protein
Gene Aliases
IKBR, NFKBIL2
Chromosome Location
Chr. 8: 144428780 - 144444488 on Build GRCh38
UniGene ID
Hs.675285
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013432.4 NP_038460.4 5 580
XM_011517050.2 XP_011515352.1 5 639
GenBank mRNA AB209126.1 - 1 167
BC008782.2 - 4 552 3628374

View Full Product Details