Gene Transcripts

Gene Symbol
OR2C1
Entrez Gene ID
4993
Gene Name
olfactory receptor family 2 subfamily C member 1
Gene Aliases
OLFmf3, OR2C2P
Chromosome Location
Chr. 16: 3355889 - 3356924 on Build GRCh38
UniGene ID
Hs.258574
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012368.2 NP_036500.2 1 325
GenBank mRNA BC069158.1 - 1 273
BC069456.1 - 1 273 7262277
BC126269.1 - 1 325 8991985
BC130328.1 - 1 325 40146359

View Full Product Details