Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014077.3 | NP_054796.1 | 2 | 140 | |
XM_011527898.1 | XP_011526200.1 | 2 | 357 | ||
GenBank mRNA | AF151902.1 | - | 2 | 99 | |
AK312058.1 | - | 2 | 123 | ||
AL050157.1 | - | 2 | 118 | ||
BC000639.1 | - | 2 | 118 | 3345441 | |
BC017286.1 | - | 2 | 101 | 4892079 | |
BG749750.1 | - | 1 | 267 |