Gene Transcripts

Gene Symbol
IL17RA
Entrez Gene ID
23765
Gene Name
interleukin 17 receptor A
Gene Aliases
CANDF5, CD217, CDw217, IL-17RA, IL17R, hIL-17R
Chromosome Location
Chr. 22: 17084959 - 17115694 on Build GRCh38
UniGene ID
Hs.48353
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289905.1 NP_001276834.1 4 471
NM_014339.6 NP_055154.3 4 471
GenBank mRNA BC011624.2 - 4 415
BX334547.2 - 4 474
BX410574.2 - 4 387
CB044543.1 - 3 454
CT841520.1 - 4 394
JF305973.1 - 4 338
U58917.1 - 4 370

View Full Product Details