Gene Transcripts

Gene Symbol
INVS
Entrez Gene ID
27130
Gene Name
inversin
Gene Aliases
INV, NPH2, NPHP2
Chromosome Location
Chr. 9: 100099185 - 100302175 on Build GRCh38
UniGene ID
Hs.558477
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318381.1 NP_001305310.1 2, 3 359
NM_001318382.1 NP_001305311.1 2, 3 359
NM_014425.4 NP_055240.2 2, 3 359
NR_134606.1 2, 3 359
GenBank mRNA AF039217.2 - 2, 3 310
AF084367.1 - 1, 2 129
AK304851.1 - 2, 3 284
AK312795.1 - 2, 3 301
BC006370.2 - 2, 3 256
BC041665.1 - 2, 3 315
BC056897.1 - 2, 3 278
BC111761.1 - 1, 2 142 40012263
BC112351.1 - 1, 2 142 40012265
BP280580.1 - 2, 3 324
DB217652.1 - 2, 3 359

View Full Product Details