Gene Transcripts

Gene Symbol
CPB2
Entrez Gene ID
1361
Gene Name
carboxypeptidase B2
Gene Aliases
CPU, PCPB, TAFI
Chromosome Location
Chr. 13: 46053186 - 46105076 on Build GRCh38
UniGene ID
Hs.512937
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278541.1 NP_001265470.1 8 794
NM_001872.4 NP_001863.3 9 905
XM_017020393.1 XP_016875882.1 9 848
GenBank mRNA AB011969.1 - 8 746
AK290829.1 - 9 853
BC007057.1 - 9 862 3934520
BT006936.1 - 9 838
M75106.1 - 9 857

View Full Product Details