Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004981.1 | NP_004972.1 | 2 | 1077 | |
NM_152868.2 | NP_690607.1 | 2 | 1239 | ||
GenBank mRNA | BC032562.1 | - | 2 | 1249 | |
BC039618.1 | - | 2 | 1122 | ||
BC113506.1 | - | 1 | 1031 | 8322558 | |
BC113508.1 | - | 1 | 1031 | 8322560 | |
CR456507.1 | - | 2 | 1047 | ||
S72503.1 | - | 2 | 1078 | ||
U07364.1 | - | 2 | 1077 | ||
U24056.1 | - | 2 | 1214 |