Gene Transcripts
- Gene Symbol
- KCTD10
- Entrez Gene ID
- 83892
- Gene Name
- potassium channel tetramerization domain containing 10
- Gene Aliases
- BTBD28, MSTP028, ULRO61, hBACURD3
- Chromosome Location
- Chr. 12: 109448655 - 109477544 on Build GRCh38
- UniGene ID
- Hs.524731
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317395.1 |
NP_001304324.1
|
5, 6 |
820 |
|
NM_031954.4 |
NP_114160.1
|
5, 6 |
817 |
|
NR_133897.1 |
|
4, 5 |
647 |
|
NR_133898.1 |
|
4, 5 |
603 |
|
GenBank mRNA |
AB209358.1 |
- |
1 |
3362 |
|
AF113208.1 |
- |
5, 6 |
562 |
|
AK027543.1 |
- |
5, 6 |
623 |
|
AK056912.1 |
- |
5, 6 |
2225 |
|
AK123733.1 |
- |
2, 3 |
1054 |
|
AK222548.1 |
- |
5, 6 |
573 |
|
AK309783.1 |
- |
4, 5 |
432 |
|
AY205299.1 |
- |
5, 6 |
623 |
|
BC040062.1 |
- |
5, 6 |
570 |
5494016 |
BX333975.2 |
- |
5, 6 |
570 |
|
CX785497.1 |
- |
4, 5 |
346 |
|
View Full Product Details