Gene Transcripts
- Gene Symbol
- MTMR7
- Entrez Gene ID
- 9108
- Gene Name
- myotubularin related protein 7
- Gene Aliases
- Chromosome Location
- Chr. 8: 17296794 - 17413531 on Build GRCh38
- UniGene ID
- Hs.625674
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004686.4 |
NP_004677.3
|
10, 11 |
1396 |
|
XM_006716414.3 |
XP_006716477.2
|
5, 6 |
1762 |
|
XM_011544692.2 |
XP_011542994.1
|
7, 8 |
835 |
|
XM_011544693.2 |
XP_011542995.1
|
6, 7 |
1297 |
|
XM_017013956.1 |
XP_016869445.1
|
9, 10 |
1085 |
|
GenBank mRNA |
AF073482.1 |
- |
6, 7 |
665 |
|
AK294468.1 |
- |
10, 11 |
1157 |
|
AK303762.1 |
- |
2, 3 |
219 |
|
AK316022.1 |
- |
10, 11 |
1206 |
|
AK316283.1 |
- |
10, 11 |
1216 |
|
BC130577.1 |
- |
10, 11 |
1182 |
40146608 |
BC130579.1 |
- |
10, 11 |
1182 |
40146610 |
CR749240.1 |
- |
10, 11 |
1194 |
|
View Full Product Details