Gene Transcripts

Gene Symbol
TREX2
Entrez Gene ID
11219
Gene Name
three prime repair exonuclease 2
Gene Aliases
Chromosome Location
Chr. X: 153444720 - 153446487 on Build GRCh38
UniGene ID
Hs.644635
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080701.3 NP_542432.2 2 639
GenBank mRNA AF151107.1 - 1 95

View Full Product Details