Gene Transcripts

Gene Symbol
USP37
Entrez Gene ID
57695
Gene Name
ubiquitin specific peptidase 37
Gene Aliases
Chromosome Location
Chr. 2: 218450251 - 218568393 on Build GRCh38
UniGene ID
Hs.166068
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020935.2 NP_065986.2 5 660
XM_005246720.2 XP_005246777.1 5 732
XM_005246721.3 XP_005246778.1 5 624
XM_005246722.3 XP_005246779.1 5 664
XM_005246724.1 XP_005246781.1 5 660
XM_011511538.2 XP_011509840.1 4 605
XM_011511541.2 XP_011509843.1 5 660
XM_017004577.1 XP_016860066.1 5 618
GenBank mRNA AB046814.1 - 1 105
AK095577.1 - 5 618
AK128583.1 - 5 664
AL832645.2 - 5 659
BC050461.1 - 5 629
BC112901.1 - 4 460 6016503
BC133007.1 - 5 545 40147029
BC133009.1 - 5 590 40147031
BC144249.1 - 5 545 9052769
BC144252.1 - 5 585 9052772
BX538024.1 - 5 625

View Full Product Details