Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014362.3 | NP_055177.2 | 14 | 1410 | |
NM_198047.2 | NP_932164.1 | 13 | 1376 | ||
XM_011510953.1 | XP_011509255.1 | 14 | 1359 | ||
XM_011510954.1 | XP_011509256.1 | 9 | 810 | ||
GenBank mRNA | AK223023.1 | - | 14 | 1134 | |
AK308042.1 | - | 14 | 1134 | ||
AK313651.1 | - | 14 | 1140 | ||
BC005190.1 | - | 13 | 1153 | ||
BC067822.1 | - | 14 | 1139 | 5266813 | |
U66669.1 | - | 14 | 1163 |