Gene Transcripts

Gene Symbol
LSM1
Entrez Gene ID
27257
Gene Name
LSM1 homolog, mRNA degradation associated
Gene Aliases
CASM, YJL124C
Chromosome Location
Chr. 8: 38163321 - 38176730 on Build GRCh38
UniGene ID
Hs.425311
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014462.2 NP_055277.1 4 1108
NR_045492.1 3 992
NR_045493.1 4 830
GenBank mRNA AF000177.1 - 4 863
AJ238094.1 - 4 886
BC001767.1 - 4 794 3353833

View Full Product Details