Gene Transcripts

Gene Symbol
OR52W1
Entrez Gene ID
120787
Gene Name
olfactory receptor family 52 subfamily W member 1
Gene Aliases
OR11-71, OR52W1P
Chromosome Location
Chr. 11: 6199224 - 6200186 on Build GRCh38
UniGene ID
Hs.553570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005178.1 NP_001005178.1 1 477
GenBank mRNA BC137225.1 - 1 555

View Full Product Details